siRNA Details
| Gene Symbol: RSPH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014433.2 | NP_055248.1 | 2, 3 | 352 | ||
| XM_011530149.2 | XP_011528451.1 | 2, 3 | 391 | |||
| XM_011530150.1 | XP_011528452.1 | 2, 3 | 391 | |||
| XM_011530151.2 | XP_011528453.1 | 2, 3 | 554 | |||
| XM_011530152.2 | XP_011528454.1 | 2, 3 | 482 | |||
| XM_017028774.1 | XP_016884263.1 | 2, 3 | 377 | |||
| GenBank mRNA | AF133587.1 | - | 2, 3 | 352 | ||
| AK299530.1 | - | 2, 3 | 310 | |||
| AK310327.1 | - | 2, 3 | 391 | |||
| BC008986.1 | - | 2, 3 | 320 | 4339021 | ||
| CR456567.1 | - | 1, 2 | 252 | |||

RSPH14
Genomic Map