siRNA Details
| Gene Symbol: EPHA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281765.2 | NP_001268694.1 | 1, 2 | 934 | ||
| NM_001281766.2 | NP_001268695.1 | 1, 2 | 934 | |||
| NM_001281767.2 | NP_001268696.1 | 1, 2 | 934 | |||
| NM_004439.7 | NP_004430.4 | 1, 2 | 934 | |||
| NM_182472.4 | NP_872272.2 | 1, 2 | 934 | |||
| XM_005265653.3 | XP_005265710.1 | 1, 2 | 1277 | |||
| XM_011531735.2 | XP_011530037.1 | 1, 2 | 1277 | |||
| XM_017007878.1 | XP_016863367.1 | 1, 2 | 1277 | |||
| XM_017007879.1 | XP_016863368.1 | 1, 2 | 1277 | |||
| XM_017007880.1 | XP_016863369.1 | 1, 2 | 1277 | |||
| XM_017007881.1 | XP_016863370.1 | 1, 2 | 1277 | |||
| GenBank mRNA | AK307826.1 | - | 1, 2 | 374 | ||
| BC136257.1 | - | 1, 2 | 201 | 9020244 | ||
| BC143427.1 | - | 1, 2 | 201 | 9051935 | ||
| L36644.1 | - | 1, 2 | 109 | |||

EPHA5
Genomic Map