siRNA Details
| Gene Symbol: NDUFA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322019.1 | NP_001308948.1 | 3 | 477 | ||
| NM_001322020.1 | NP_001308949.1 | 3 | 477 | |||
| NM_004544.3 | NP_004535.1 | 3 | 477 | |||
| NR_136155.1 | 3 | 477 | ||||
| NR_136156.1 | 3 | 477 | ||||
| NR_136157.1 | 2 | 308 | ||||
| NR_136158.1 | 3 | 477 | ||||
| XM_011511228.2 | XP_011509530.1 | 3 | 464 | |||
| GenBank mRNA | AB209440.1 | - | 3 | 398 | ||
| AF087661.1 | - | 3 | 397 | |||
| AF087854.1 | - | 3 | 416 | |||
| AF453834.1 | - | 3 | 376 | |||
| AK290778.1 | - | 3 | 399 | |||
| BC003417.1 | - | 3 | 411 | 3451514 | ||
| BC031332.1 | - | 3 | 382 | 4872196 | ||
| BI551914.1 | - | 461 | ||||

NDUFA10
Genomic Map