siRNA Details
| Gene Symbol: CDK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261435.1 | NP_001248364.1 | 4 | 511 | ||
| NM_001261436.1 | NP_001248365.1 | 4 | 511 | |||
| NM_139158.2 | NP_631897.1 | 4 | 421 | |||
| XM_005246781.3 | XP_005246838.1 | 4 | 511 | |||
| XM_005246782.4 | XP_005246839.1 | 5 | 758 | |||
| XM_011511650.2 | XP_011509952.1 | 4 | 511 | |||
| XM_011511652.1 | XP_011509954.1 | 4 | 511 | |||
| XM_011511654.2 | XP_011509956.1 | 4 | 511 | |||
| GenBank mRNA | AB053308.1 | - | 4 | 375 | ||
| AK131512.1 | - | 4 | 511 | |||
| AK292434.1 | - | 5 | 628 | |||
| AK310050.1 | - | 6 | 744 | |||
| AX772791.1 | - | 4 | 375 | |||
| BC038807.1 | - | 4 | 469 | 5200755 | ||
| BX647394.1 | - | 4 | 475 | |||
| DA050598.1 | - | 4 | 511 | |||

CDK15
Genomic Map