siRNA Details
| Gene Symbol: GPR155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033045.3 | NP_001028217.1 | 3 | 507 | ||
| NM_001267050.1 | NP_001253979.1 | 3 | 404 | |||
| NM_001267051.1 | NP_001253980.1 | 2 | 358 | |||
| NM_152529.6 | NP_689742.4 | 2 | 358 | |||
| XM_017003485.1 | XP_016858974.1 | 4 | 549 | |||
| XM_017003486.1 | XP_016858975.1 | 2 | 410 | |||
| XM_017003487.1 | XP_016858976.1 | 3 | 411 | |||
| GenBank mRNA | AK091200.1 | - | 2 | 299 | ||
| AK307896.1 | - | 2 | 350 | |||
| AK315125.1 | - | 2 | 299 | |||
| AL834154.1 | - | 3 | 456 | |||
| BC028730.1 | - | 3 | 453 | 4813561 | ||
| BC035037.1 | - | 3 | 415 | 5260361 | ||
| BC036681.1 | - | 2 | 304 | |||
| BI548910.1 | - | 3 | 415 | |||
| BX537947.1 | - | 2 | 351 | |||
| DC334438.1 | - | 2 | 358 | |||

GPR155
Genomic Map