siRNA Details
| Gene Symbol: TMEM171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161342.2 | NP_001154814.1 | 2 | 812 | ||
| NM_173490.7 | NP_775761.4 | 2 | 812 | |||
| XM_011543156.1 | XP_011541458.1 | 2 | 792 | |||
| GenBank mRNA | BC018083.1 | - | 2 | 798 | 4797244 | |
| BC035310.1 | - | 1 | 653 | 4746772 | ||

TMEM171
Genomic Map