siRNA Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003712.3 | NP_003703.1 | 6 | 1240 | ||
| NM_177526.2 | NP_803545.1 | 6 | 1199 | |||
| NM_177543.2 | NP_808211.1 | 6 | 1307 | |||
| XM_011528396.2 | XP_011526698.1 | 6 | 1236 | |||
| GenBank mRNA | AB209696.1 | - | 3 | 6388 | ||
| AF035959.1 | - | 6 | 1182 | |||
| BC002806.2 | - | 6 | 1191 | 3659728 | ||

PLPP2
Genomic Map