siRNA Details
| Gene Symbol: NDUFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004541.3 | NP_004532.1 | 3 | 367 | ||
| GenBank mRNA | BC000266.1 | - | 3 | 264 | 3352028 | |
| BQ680674.1 | - | 3 | 268 | |||
| U54993.1 | - | 3 | 259 | |||
| X81900.1 | - | 3 | 243 | |||

NDUFA1
Genomic Map