siRNA Details
| Gene Symbol: ASB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142459.1 | NP_001135931.2 | 6 | 1909 | ||
| NM_001142460.1 | NP_001135932.2 | 5 | 1795 | |||
| NM_080871.3 | NP_543147.2 | 6 | 1713 | |||
| GenBank mRNA | AF417920.1 | - | 5 | 1797 | ||
| AK055536.1 | - | 6 | 1619 | |||
| AK125565.1 | - | 6 | 1713 | |||

ASB10
Genomic Map