siRNA Details
| Gene Symbol: TMEM86B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173804.4 | NP_776165.3 | 1 | 117 | ||
| GenBank mRNA | BC023000.1 | - | 1 | 117 | 5000011 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM86B| Gene Symbol: TMEM86B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173804.4 | NP_776165.3 | 1 | 117 | ||
| GenBank mRNA | BC023000.1 | - | 1 | 117 | 5000011 | |