siRNA Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002793.3 | NP_002784.1 | 5, 6 | 632 | ||
| GenBank mRNA | AB451312.1 | - | 5, 6 | 545 | ||
| AB451442.1 | - | 5, 6 | 545 | |||
| AK023290.1 | - | 4, 5 | 1760 | |||
| AK058050.1 | - | 5, 6 | 602 | |||
| AK223193.1 | - | 5, 6 | 583 | |||
| BC000508.2 | - | 5, 6 | 583 | 2822268 | ||
| BC020807.1 | - | 5, 6 | 583 | 4281426 | ||
| BQ685374.1 | - | 4, 5 | 362 | |||
| BT019720.1 | - | 5, 6 | 545 | |||
| D00761.1 | - | 5, 6 | 557 | |||

PSMB1
Genomic Map