siRNA Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320108.1 | NP_001307037.1 | 2 | 2302 | ||
| NM_003708.4 | NP_003699.3 | 3 | 2561 | |||
| GenBank mRNA | AB209332.1 | - | 2 | 2302 | ||
| AF057034.1 | - | 3 | 785 | |||
| AF086735.1 | - | 3 | 1501 | |||

RDH16
Genomic Map