siRNA Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004419.3 | NP_004410.3 | 3, 4 | 996 | ||
| GenBank mRNA | AB209823.1 | - | 3, 4 | 942 | ||
| BC062545.1 | - | 3, 4 | 913 | 5808876 | ||
| U15932.2 | - | 3, 4 | 948 | |||
| U16996.1 | - | 3, 4 | 787 | |||
| Gene Symbol: DUSP5P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002834.1 | 2 | 1025 | |||
| GenBank mRNA | AK092260.1 | - | 2 | 1025 | ||

DUSP5
Genomic Map