siRNA Details
Gene Symbol: OR1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012353.2 | NP_036485.2 | 1 | 815 | ||
GenBank mRNA | BC140914.1 | - | 1 | 1087 | 9056726 | |
X89674.1 | - | 1 | 441 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012353.2 | NP_036485.2 | 1 | 815 | ||
GenBank mRNA | BC140914.1 | - | 1 | 1087 | 9056726 | |
X89674.1 | - | 1 | 441 |