siRNA Details
| Gene Symbol: CNTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243270.1 | NP_001230199.1 | 5 | 871 | ||
| NM_001243271.1 | NP_001230200.1 | 6 | 1010 | |||
| NM_014361.3 | NP_055176.1 | 6 | 1010 | |||
| NM_175566.2 | NP_780775.1 | 3 | 354 | |||
| XM_011542871.1 | XP_011541173.1 | 5 | 788 | |||
| XM_011542873.1 | XP_011541175.1 | 6 | 1010 | |||
| XM_017017926.1 | XP_016873415.1 | 7 | 1178 | |||
| XM_017017927.1 | XP_016873416.1 | 7 | 1178 | |||
| XM_017017928.1 | XP_016873417.1 | 5 | 943 | |||
| XM_017017929.1 | XP_016873418.1 | 5 | 788 | |||
| GenBank mRNA | AB013802.2 | - | 4 | 585 | ||
| AB013803.2 | - | 3 | 363 | |||
| AK308788.1 | - | 6 | 1010 | |||
| BC039255.1 | - | 6 | 845 | 5273769 | ||
| BC130619.1 | - | 5 | 657 | 40146650 | ||
| BC144185.1 | - | 5 | 657 | 9052705 | ||
| BC144186.1 | - | 6 | 796 | 9052706 | ||

CNTN5
Genomic Map