siRNA Details
| Gene Symbol: OR10A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003745.1 | NP_001003745.1 | 1 | 234 | ||
| GenBank mRNA | BC137031.1 | - | 1 | 308 | 9021027 | |
| BC137032.1 | - | 1 | 308 | 9021028 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10A3| Gene Symbol: OR10A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003745.1 | NP_001003745.1 | 1 | 234 | ||
| GenBank mRNA | BC137031.1 | - | 1 | 308 | 9021027 | |
| BC137032.1 | - | 1 | 308 | 9021028 | ||