siRNA Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258.2 | NP_001249.1 | 7 | 714 | ||
GenBank mRNA | X66357.1 | - | 6 | 726 |
Gene Symbol: TEN1-CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037709.1 | 9 | 2474 | |||
GenBank mRNA | AK097104.1 | - | 9 | 2431 | ||
BX647274.1 | - | 8 | 3042 |
