siRNA Details
| Gene Symbol: UBE2G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 2102 | ||
| NM_003343.5 | NP_003334.2 | 6 | 2184 | |||
| NM_182688.2 | NP_872630.1 | 7 | 2299 | |||
| XM_017028469.1 | XP_016883958.1 | 6 | 2143 | |||
| GenBank mRNA | AF032456.1 | - | 6 | 2151 | ||
| AK122700.1 | - | 2 | 3601 | |||
| AL834129.1 | - | 2 | 2920 | |||
| BC001738.1 | - | 6 | 2147 | 3536853 | ||
| BC008351.1 | - | 6 | 2144 | 3536171 | ||

UBE2G2
Genomic Map