siRNA Details
| Gene Symbol: RRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006583.2 | NP_006574.1 | 2, 3 | 331 | ||
| GenBank mRNA | AF012270.1 | - | 2, 3 | 347 | ||
| BC128120.1 | - | 2, 3 | 322 | 40112275 | ||
| BC128121.1 | - | 2, 3 | 322 | 40112276 | ||
| BC128401.1 | - | 2, 3 | 322 | 40112297 | ||
More Information
Set Membership: |
|

RRH
Genomic Map