siRNA Details
| Gene Symbol: CPNE7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014427.4 | NP_055242.1 | 6, 7 | 870 | ||
| NM_153636.2 | NP_705900.1 | 4, 5 | 645 | |||
| XM_011523000.1 | XP_011521302.1 | 4, 5 | 640 | |||
| XM_011523001.2 | XP_011521303.1 | 4, 5 | 405 | |||
| XM_017023138.1 | XP_016878627.1 | 6, 7 | 834 | |||
| XM_017023139.1 | XP_016878628.1 | 6, 7 | 845 | |||
| XM_017023140.1 | XP_016878629.1 | 4, 5 | 620 | |||
| XM_017023141.1 | XP_016878630.1 | 3, 4 | 1727 | |||
| GenBank mRNA | AJ133798.1 | - | 6, 7 | 869 | ||
| AL539371.3 | - | 4, 5 | 647 | |||
| BC035334.1 | - | 4, 5 | 583 | 5191900 | ||
| BC064577.1 | - | 4, 5 | 622 | 5727324 | ||

CPNE7
Genomic Map