siRNA Details
| Gene Symbol: MORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014429.3 | NP_055244.3 | 6 | 455 | ||
| XM_005247362.2 | XP_005247419.1 | 6 | 412 | |||
| XM_011512691.1 | XP_011510993.1 | 6 | 412 | |||
| XM_011512692.1 | XP_011510994.1 | 6 | 412 | |||
| XM_011512693.1 | XP_011510995.1 | 6 | 412 | |||
| XM_011512694.1 | XP_011510996.1 | 3 | 163 | |||
| XM_017006169.1 | XP_016861658.1 | 6 | 412 | |||
| GenBank mRNA | AF084946.1 | - | 6 | 430 | ||
| AK302642.1 | - | 6 | 412 | |||
| BC028680.1 | - | 6 | 456 | |||
| BC050307.1 | - | 6 | 394 | 4838254 | ||

MORC1
Genomic Map