siRNA Details
| Gene Symbol: NAV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024383.1 | NP_001019554.1 | 13 | 2874 | ||
| NM_014903.5 | NP_055718.4 | 13 | 2874 | |||
| XM_005269215.3 | XP_005269272.1 | 13 | 6875 | |||
| XM_011538940.2 | XP_011537242.1 | 13 | 6876 | |||
| XM_011538941.2 | XP_011537243.1 | 13 | 6875 | |||
| XM_011538944.2 | XP_011537246.1 | 13 | 6873 | |||
| XM_017020164.1 | XP_016875653.1 | 13 | 6877 | |||
| XM_017020165.1 | XP_016875654.1 | 13 | 6877 | |||
| XM_017020166.1 | XP_016875655.1 | 13 | 2863 | |||
| XM_017020167.1 | XP_016875656.1 | 12 | 2794 | |||
| XM_017020168.1 | XP_016875657.1 | 13 | 6876 | |||
| XM_017020169.1 | XP_016875658.1 | 13 | 6875 | |||
| XM_017020170.1 | XP_016875659.1 | 13 | 6872 | |||
| XM_017020171.1 | XP_016875660.1 | 5 | 6170 | |||
| XM_017020172.1 | XP_016875661.1 | 5 | 1808 | |||
| XM_017020173.1 | XP_016875662.1 | 5 | 6168 | |||
| XM_017020174.1 | XP_016875663.1 | 5 | 6168 | |||
| XM_017020175.1 | XP_016875664.1 | 5 | 2047 | |||
| GenBank mRNA | AF397731.1 | - | 12 | 2458 | ||
| AJ488201.1 | - | 13 | 2746 | |||

NAV3
Genomic Map