siRNA Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278945.1 | NP_001265874.1 | 5 | 832 | ||
| NM_018492.3 | NP_060962.2 | 5 | 890 | |||
| XM_006716368.2 | XP_006716431.1 | 5 | 814 | |||
| GenBank mRNA | AB027249.1 | - | 5 | 616 | ||
| AB027250.1 | - | 5 | 596 | |||
| AF189722.1 | - | 5 | 580 | |||
| AF237709.1 | - | 5 | 604 | |||
| AK027291.1 | - | 5 | 595 | |||
| AK301836.1 | - | 5 | 792 | |||
| BC015191.1 | - | 5 | 604 | 4082846 | ||
| EU794685.1 | - | 5 | 627 | |||

PBK
Genomic Map