siRNA Details
| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317987.1 | NP_001304916.1 | 2 | 273 | ||
| NM_014443.2 | NP_055258.1 | 2 | 249 | |||
| XM_011537622.2 | XP_011535924.1 | 3 | 572 | |||
| XM_017009346.1 | XP_016864835.1 | 2 | 766 | |||
| XM_017009347.1 | XP_016864836.1 | 4 | 480 | |||
| XM_017009348.1 | XP_016864837.1 | 3 | 363 | |||
| GenBank mRNA | AF110385.1 | - | 2 | 242 | ||
| AF152098.1 | - | 2 | 240 | |||
| AF184969.1 | - | 2 | 240 | |||
| AF212311.1 | - | 2 | 249 | |||
| AF218727.1 | - | 2 | 240 | |||
| AY358931.1 | - | 2 | 240 | |||
| BC113925.1 | - | 2 | 222 | 40084832 | ||
| BC113946.1 | - | 2 | 222 | 40084836 | ||
| BM826445.1 | - | 2 | 273 | |||
| CR457192.1 | - | 2 | 199 | |||

IL17B
Genomic Map