siRNA Details
| Gene Symbol: ITPKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002220.2 | NP_002211.1 | 4 | 936 | ||
| XM_011521522.2 | XP_011519824.1 | 5 | 617 | |||
| GenBank mRNA | BC026331.1 | - | 4 | 920 | 4792595 | |
| X54938.1 | - | 4 | 877 | |||

ITPKA
Genomic Map