siRNA Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289905.1 | NP_001276834.1 | 12 | 1176 | ||
| NM_014339.6 | NP_055154.3 | 13 | 1278 | |||
| GenBank mRNA | BC011624.2 | - | 13 | 1222 | 4053746 | |
| BX368715.2 | - | 6 | 347 | |||
| CT841520.1 | - | 13 | 1201 | |||
| JF305973.1 | - | 13 | 1145 | |||
| U58917.1 | - | 13 | 1177 | |||

IL17RA
Genomic Map