siRNA Details
| Gene Symbol: ZNF324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014347.2 | NP_055162.1 | 4 | 2673 | ||
| XM_005258713.4 | XP_005258770.1 | 4 | 3468 | |||
| GenBank mRNA | AF060503.1 | - | 4 | 2622 | ||
| AF075117.1 | - | 1 | 190 | |||
| AK023989.1 | - | 2 | 1936 | |||
| AK092341.1 | - | 4 | 3024 | |||
| BC007717.2 | - | 5 | 2513 | 4303415 | ||

ZNF324
Genomic Map