siRNA Details
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 4 | 2159 | ||
NM_206852.2 | NP_996734.1 | 2 | 554 | |||
XM_017021578.1 | XP_016877067.1 | 2 | 443 | |||
GenBank mRNA | AK290534.1 | - | 2 | 554 | ||
AK297492.1 | - | 3 | 461 | |||
BC000314.2 | - | 2 | 339 | |||
BC003003.1 | - | 2 | 339 | |||
BC090862.1 | - | 4 | 2074 | 6179803 | ||
BC111694.1 | - | 5 | 1849 | 40035553 | ||
BX449967.2 | - | 2 | 542 | |||
CR457030.1 | - | 2 | 246 | |||
L10333.1 | - | 4 | 2072 | |||
L10334.1 | - | 2 | 980 | |||
L10335.1 | - | 2 | 274 |
