siRNA Details
| Gene Symbol: MS4A6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001247999.1 | NP_001234928.1 | 3 | 691 | ||
| NM_022349.3 | NP_071744.2 | 2 | 502 | |||
| NM_152851.2 | NP_690590.1 | 2 | 502 | |||
| NM_152852.2 | NP_690591.1 | 3 | 627 | |||
| XM_005274177.2 | XP_005274234.1 | 3 | 686 | |||
| XM_006718658.2 | XP_006718721.1 | 3 | 561 | |||
| XM_006718660.2 | XP_006718723.1 | 3 | 561 | |||
| XM_006718661.2 | XP_006718724.1 | 3 | 561 | |||
| XM_011545209.1 | XP_011543511.1 | 3 | 561 | |||
| XM_017018123.1 | XP_016873612.1 | 2 | 297 | |||
| XM_017018124.1 | XP_016873613.1 | 2 | 297 | |||
| XM_017018125.1 | XP_016873614.1 | 3 | 561 | |||
| GenBank mRNA | AB013104.1 | - | 2 | 210 | ||
| AF142409.1 | - | 2 | 230 | |||
| AF172819.1 | - | 3 | 648 | |||
| AF212240.1 | - | 2 | 371 | |||
| AF253977.1 | - | 2 | 372 | |||
| AF261136.1 | - | 2 | 369 | |||
| AF350502.1 | - | 2 | 219 | |||
| AF350503.1 | - | 2 | 219 | |||
| AK026454.1 | - | 2 | 325 | |||
| AK291870.1 | - | 3 | 627 | |||
| AK300725.1 | - | 2 | 236 | |||
| AK301387.1 | - | 2 | 197 | |||
| BC022854.1 | - | 2 | 241 | 3934920 | ||
| BC108728.1 | - | 2 | 275 | 6196213 | ||
| DA917243.1 | - | 2 | 326 | |||
| DA979138.1 | - | 2 | 273 | |||

MS4A6A
Genomic Map