siRNA Details
| Gene Symbol: ENSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004436.2 | NP_004427.1 | 2 | 288 | ||
| NM_207042.1 | NP_996925.1 | 2 | 288 | |||
| NM_207043.1 | NP_996926.1 | 2 | 288 | |||
| NM_207044.1 | NP_996927.1 | 2 | 288 | |||
| NM_207045.1 | NP_996928.1 | 2 | 152 | |||
| NM_207046.1 | NP_996929.1 | 2 | 152 | |||
| NM_207047.1 | NP_996930.1 | 2 | 152 | |||
| NM_207168.1 | NP_997051.1 | 2 | 288 | |||
| GenBank mRNA | AF067170.1 | - | 2 | 245 | ||
| AF157509.1 | - | 2 | 242 | |||
| AF157510.1 | - | 2 | 242 | |||
| AK001981.1 | - | 2 | 262 | |||
| AK057673.1 | - | 1 | 2084 | |||
| AK290064.1 | - | 2 | 135 | |||
| AL711949.1 | - | 2 | 291 | |||
| BC000436.2 | - | 2 | 236 | 2820528 | ||
| BC004461.1 | - | 2 | 236 | 2820528 | ||
| BC068544.1 | - | 2 | 253 | 5273162 | ||
| BC069208.1 | - | 2 | 226 | 4853917 | ||
| BE799496.1 | - | 2 | 172 | |||
| BQ883543.1 | - | 2 | 250 | |||
| BU075511.1 | - | 2 | 213 | |||
| BU858280.1 | - | 146 | ||||
| CR457130.1 | - | 2 | 136 | |||
| CR536578.1 | - | 2 | 136 | |||
| CR749580.1 | - | 2 | 243 | |||
| T78564.1 | - | 2 | 167 | |||
| X99906.1 | - | 2 | 261 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019561, NM_001026212, BC059724, AK149726, AK006149, and others.

ENSA
Genomic Map