siRNA Details
| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF222904.1 | - | 2 | 811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYY2| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF222904.1 | - | 2 | 811 | ||