siRNA Details
| Gene Symbol: OXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000915.3 | NP_000906.1 | 3 | 423 | ||
| XM_011529238.1 | XP_011527540.1 | 4 | 560 | |||
| GenBank mRNA | BC069144.1 | - | 3 | 423 | 7217004 | |
| BC101841.1 | - | 3 | 421 | 8069347 | ||
| BC101843.1 | - | 3 | 421 | 8069349 | ||
| M25650.1 | - | 3 | 404 | |||
| X03173.1 | - | 2 | 151 | |||

OXT
Genomic Map