siRNA Details
| Gene Symbol: Kcna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012971.2 | NP_037103.1 | 1 | 2664 | ||
| GenBank mRNA | M32867.1 | - | 1 | 2664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna4| Gene Symbol: Kcna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012971.2 | NP_037103.1 | 1 | 2664 | ||
| GenBank mRNA | M32867.1 | - | 1 | 2664 | ||