siRNA Details
| Gene Symbol: Kcna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012971.2 | NP_037103.1 | 1 | 2760 | ||
| XM_006234673.2 | XP_006234735.1 | 2 | 3961 | |||
| XM_006234674.2 | XP_006234736.1 | 2 | 3813 | |||
| GenBank mRNA | M32867.1 | - | 1 | 2760 | ||
| X16002.1 | - | 1 | 3173 | |||

Kcna4
Genomic Map