siRNA Details
| Gene Symbol: Slc17a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053427.1 | NP_445879.1 | 7 | 1298 | ||
| XM_006229265.1 | XP_006229327.1 | 7 | 827 | |||
| GenBank mRNA | AF271235.1 | - | 7 | 1298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a6| Gene Symbol: Slc17a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053427.1 | NP_445879.1 | 7 | 1298 | ||
| XM_006229265.1 | XP_006229327.1 | 7 | 827 | |||
| GenBank mRNA | AF271235.1 | - | 7 | 1298 | ||