siRNA Details
| Gene Symbol: Kcnq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133322.1 | NP_579856.1 | 16 | 2675 | ||
| GenBank mRNA | AF087453.2 | - | 16 | 2675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq2| Gene Symbol: Kcnq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133322.1 | NP_579856.1 | 16 | 2675 | ||
| GenBank mRNA | AF087453.2 | - | 16 | 2675 | ||