siRNA Details
Gene Symbol: GPR146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303473.1 | NP_001290402.1 | 2 | 1696 | ||
NM_001303474.1 | NP_001290403.1 | 3 | 1964 | |||
NM_138445.3 | NP_612454.1 | 2 | 1819 | |||
XM_005249613.2 | XP_005249670.1 | 3 | 2373 | |||
XM_006715642.2 | XP_006715705.1 | 3 | 2541 | |||
XM_006715643.2 | XP_006715706.1 | 3 | 2491 | |||
XM_011515116.2 | XP_011513418.1 | 3 | 1938 | |||
GenBank mRNA | AK300166.1 | - | 2 | 1402 | ||
BC014241.2 | - | 1 | 1597 | 4563636 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
