siRNA Details
| Gene Symbol: CSNK1A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145203.5 | NP_660204.2 | 1 | 2367 | ||
| GenBank mRNA | BC028723.2 | - | 1 | 2369 | 5266487 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK1A1L| Gene Symbol: CSNK1A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145203.5 | NP_660204.2 | 1 | 2367 | ||
| GenBank mRNA | BC028723.2 | - | 1 | 2369 | 5266487 | |