siRNA Details
| Gene Symbol: TAS2R8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023918.1 | NP_076407.1 | 1 | 93 | ||
| GenBank mRNA | BC069084.1 | - | 1 | 93 | ||
| BC069551.1 | - | 1 | 93 | 7262241 | ||
| BC096701.1 | - | 1 | 93 | 40015728 | ||
| BC096735.1 | - | 1 | 93 | 40015730 | ||
| BC098167.1 | - | 1 | 93 | 40015729 | ||
| BC098353.1 | - | 1 | 93 | 40015727 | ||

TAS2R8
Genomic Map