siRNA Details
| Gene Symbol: DYNC1LI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016141.3 | NP_057225.2 | 3, 4 | 444 | ||
| GenBank mRNA | AF078849.1 | - | 3, 4 | 420 | ||
| AK074348.1 | - | 3, 4 | 428 | |||
| AK222573.1 | - | 3, 4 | 379 | |||
| AK222653.1 | - | 3, 4 | 380 | |||
| BC131620.1 | - | 3, 4 | 390 | 40117391 | ||
| BX649189.1 | - | 3, 4 | 263 | |||
| DA284201.1 | - | 3, 4 | 444 | |||
| HY036998.1 | - | 3, 4 | 497 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146229, AK180961, BC129879, BC023347, AK159702.

DYNC1LI1
Genomic Map