siRNA Details
| Gene Symbol: PPIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032472.3 | NP_115861.1 | 6 | 585 | ||
| NM_130906.2 | NP_570981.1 | 6 | 573 | |||
| XM_005246651.3 | XP_005246708.1 | 6 | 504 | |||
| XM_005246652.4 | XP_005246709.1 | 6 | 492 | |||
| XM_011511358.2 | XP_011509660.1 | 6 | 678 | |||
| XM_017004352.1 | XP_016859841.1 | 6 | 695 | |||
| XM_017004353.1 | XP_016859842.1 | 6 | 434 | |||
| XM_017004354.1 | XP_016859843.1 | 6 | 868 | |||
| XM_017004355.1 | XP_016859844.1 | 6 | 892 | |||
| XM_017004356.1 | XP_016859845.1 | 6 | 856 | |||
| XM_017004357.1 | XP_016859846.1 | 7 | 775 | |||
| XM_017004358.1 | XP_016859847.1 | 7 | 598 | |||
| XM_017004359.1 | XP_016859848.1 | 7 | 782 | |||
| XM_017004360.1 | XP_016859849.1 | 5 | 595 | |||
| GenBank mRNA | AF146799.1 | - | 6 | 442 | ||
| AF251049.1 | - | 6 | 585 | |||
| AF271652.1 | - | 6 | 508 | |||
| AF414127.1 | - | 5 | 509 | |||
| AK297804.1 | - | 6 | 504 | |||
| AY257468.1 | - | 5 | 629 | |||
| BC007693.2 | - | 6 | 1014 | 3956681 | ||
| CB961908.1 | - | 6 | 628 | |||
| KU178646.1 | - | 4 | 332 | |||
| KU178647.1 | - | 5 | 385 | |||
| KU178648.1 | - | 4 | 344 | |||

PPIL3
Genomic Map