siRNA Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301409.1 | NP_001288338.1 | 6 | 1096 | ||
| NM_198141.2 | NP_937784.2 | 5 | 709 | |||
| GenBank mRNA | AF545045.1 | - | 5 | 403 | ||
| AF545046.1 | - | 5 | 403 | |||
| AK074037.1 | - | 5 | 601 | |||
| AK074330.1 | - | 6 | 1098 | |||
| BC059406.1 | - | 5 | 581 | 30330796 | ||
| BC093833.1 | - | 5 | 426 | 7939678 | ||
| BC112051.1 | - | 5 | 426 | 8327519 | ||

GANC
Genomic Map