siRNA Details
| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176958.3 | NP_795932.2 | 2 | 293 | ||
| GenBank mRNA | AK043636.1 | - | 2 | 279 | ||
| AK084960.1 | - | 2 | 288 | |||
| AK141867.1 | - | 2 | 279 | |||
| AK154774.1 | - | 2 | 293 | |||
| BC130013.1 | - | 2 | 297 | 40140349 | ||
| BY122345.1 | - | 2 | 297 | |||
| BY713373.1 | - | 2 | 279 | |||

Hif1an
Genomic Map