siRNA Details
| Gene Symbol: PALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014431.2 | NP_055246.2 | 11 | 1560 | ||
| XM_011539637.2 | XP_011537939.1 | 11 | 2746 | |||
| XM_011539638.2 | XP_011537940.1 | 11 | 1333 | |||
| XM_017016072.1 | XP_016871561.1 | 11 | 1331 | |||
| GenBank mRNA | AB033100.2 | - | 11 | 1558 | ||
| BC040163.1 | - | 9 | 1063 | |||
| BC136375.1 | - | 11 | 1442 | 9020362 | ||
| BC136376.1 | - | 11 | 1442 | 9020363 | ||

PALD1
Genomic Map