siRNA Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243750.1 | NP_001230679.1 | 4 | 732 | ||
| NM_181843.2 | NP_862826.1 | 3 | 803 | |||
| GenBank mRNA | AK123561.1 | - | 4 | 716 | ||
| BC018644.1 | - | 3 | 796 | 3956297 | ||

NUDT8
Genomic Map