siRNA Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181843.2 | NP_862826.1 | 3 | 806 | ||
| GenBank mRNA | BC018644.1 | - | 3 | 799 | 3956297 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUDT8| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181843.2 | NP_862826.1 | 3 | 806 | ||
| GenBank mRNA | BC018644.1 | - | 3 | 799 | 3956297 | |