siRNA Details
| Gene Symbol: ARSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000047.2 | NP_000038.2 | 10 | 1608 | ||
| NM_001282628.1 | NP_001269557.1 | 11 | 1894 | |||
| NM_001282631.1 | NP_001269560.1 | 9 | 1378 | |||
| XM_005274518.2 | XP_005274575.1 | 10 | 1535 | |||
| XM_005274519.4 | XP_005274576.1 | 10 | 1698 | |||
| XM_005274521.4 | XP_005274578.1 | 9 | 1491 | |||
| XM_011545521.1 | XP_011543823.1 | 8 | 1178 | |||
| XM_017029525.1 | XP_016885014.1 | 11 | 1825 | |||
| XM_017029526.1 | XP_016885015.1 | 9 | 1378 | |||
| GenBank mRNA | AK223183.1 | - | 10 | 1438 | ||
| AK223199.1 | - | 10 | 1405 | |||
| AK293648.1 | - | 10 | 1313 | |||
| AK301004.1 | - | 11 | 1725 | |||
| AK313093.1 | - | 10 | 1427 | |||
| BC130438.1 | - | 10 | 1403 | 40146469 | ||
| X83573.1 | - | 10 | 1425 | |||

ARSE
Genomic Map