siRNA Details
| Gene Symbol: PLA2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007267.2 | NP_001007268.1 | 4, 5 | 1063 | ||
| NM_001195641.1 | NP_001182570.1 | 4, 5 | 1063 | |||
| NM_007366.4 | NP_031392.3 | 4, 5 | 1063 | |||
| XM_005246392.3 | XP_005246449.1 | 4, 5 | 951 | |||
| XM_011510820.2 | XP_011509122.1 | 4, 5 | 951 | |||
| XM_017003598.1 | XP_016859087.1 | 4, 5 | 951 | |||
| XM_017003599.1 | XP_016859088.1 | 4, 5 | 951 | |||
| XM_017003601.1 | XP_016859090.1 | 4, 5 | 1350 | |||
| GenBank mRNA | BC140823.1 | - | 4, 5 | 873 | 9021691 | |
| BC144631.1 | - | 4, 5 | 873 | 9053162 | ||
| U17033.1 | - | 4, 5 | 1063 | |||
| U17034.1 | - | 4, 5 | 1063 | |||

PLA2R1
Genomic Map