siRNA Details
| Gene Symbol: PIP5K1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003558.3 | NP_003549.1 | 15 | 2106 | ||
| XM_005252261.3 | XP_005252318.1 | 14 | 2146 | |||
| XM_005252262.4 | XP_005252319.1 | 15 | 1902 | |||
| XM_006717300.2 | XP_006717363.1 | 15 | 2012 | |||
| XM_011519082.2 | XP_011517384.1 | 15 | 4773 | |||
| XM_011519083.2 | XP_011517385.1 | 15 | 1927 | |||
| XM_011519084.2 | XP_011517386.1 | 16 | 2046 | |||
| XM_017015188.1 | XP_016870677.1 | 15 | 1956 | |||
| XM_017015189.1 | XP_016870678.1 | 16 | 2071 | |||
| XM_017015190.1 | XP_016870679.1 | 14 | 1892 | |||
| GenBank mRNA | AK292734.1 | - | 15 | 1823 | ||
| BC030587.2 | - | 20 | 2463 | 4823087 | ||
| U78579.1 | - | 8 | 897 | |||
| U78580.1 | - | 6 | 498 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008846, AK149354, AK164552, AF048695, D86176, and others.

PIP5K1B
Genomic Map