siRNA Details
| Gene Symbol: DIRAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004675.3 | NP_004666.1 | 4 | 1385 | ||
| XM_017002773.1 | XP_016858262.1 | 2 | 2028 | |||
| GenBank mRNA | AK021882.1 | - | 1 | 1152 | ||
| AK096393.1 | - | 4 | 1385 | |||
| BC005362.1 | - | 2 | 895 | 3686616 | ||
| U96750.1 | - | 2 | 896 | |||

DIRAS3
Genomic Map